Learn More
Biomatik Corporation Human Deoxyribonuclease I Like Protein 3 (DNASE1L3) ELISA Kit
Supplier:  Biomatik Corporation EKU10790
Encompass_Preferred
Human Deoxyribonuclease I Like Protein 3 (DNASE1L3) ELISA Kit for detecting DNASE1L3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13609. Gene ID: 1776. Target Synonyms: DHP2; DNAS1L3; LSD; Deoxyribonuclease Gamma; Liver and spleen DNase; DNase I homolog protein DHP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6766