Learn More
Biomatik Corporation Human Prokineticin 2 (PK2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06814
Encompass_Preferred
The Human Prokineticin 2 (PK2) ELISA Kit allows for quantitative determination of Prokineticin 2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 246.91-20000 pg/mL. Sensitivity: 95.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q9HC23. Cross Activity: No significant cross-reactivity or interference between Prokineticin 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3608