Learn More
Biomatik Corporation Human Lipoprotein Α, Lp-Α Elisa Kit
Supplier:  Biomatik Corporation EKC31272
Encompass_Preferred
Human Lipoprotein Α, Lp-Α Elisa Kit for detecting Lipoprotein, Lp(A) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000ng/ml. Sensitivity: 53.106ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Target Synonyms: AK38; APOA; LP; OTTHUMP00000017543; antiangiogenic AK38 protein; apolipoprotein (a); lipoprotein Lp (a). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7166