Learn More
Biomatik Corporation Human Protein C Receptor, Endothelial (PROCR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06896
Encompass_Preferred
The Human Protein C Receptor, Endothelial (PROCR) ELISA Kit allows for quantitative determination of Protein C Receptor, Endothelial in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.269 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UNN8. Cross Activity: No significant cross-reactivity or interference between Protein C Receptor, Endothelial and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3636