Learn More
Biomatik Corporation Canine Thymus Activation Regulated Chemokine (Tarc/Ccl17) Elisa Kit
Supplier:  Biomatik Corporation EKC31885
Encompass_Preferred
Canine Thymus Activation Regulated Chemokine (Tarc/Ccl17) Elisa Kit for detecting Thymus Activation Regulated Chemokine(Tarc/Ccl17) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q95N01; CCL17. Target Synonyms: CCL17; TARCC-C motif chemokine 17; CC chemokine TARC; Small-inducible cytokine A17; Thymus and activation-regulated chemokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7236