Learn More
Biomatik Corporation Chicken Interleukin 2, Il-2 Elisa Kit
Supplier:  Biomatik Corporation EKC31956
Encompass_Preferred
Chicken Interleukin 2, Il-2 Elisa Kit for detecting Interleukin 2, Il-2 in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 0.18-12pg/ml. Sensitivity: 0.04pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O73883. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7245