Learn More
Biomatik Corporation Human Bone Alkaline Phosphatase, Balp Elisa Kit
Supplier:  Biomatik Corporation EKC32886
Encompass_Preferred
Human Bone Alkaline Phosphatase, Balp Elisa Kit for detecting Bone Alkaline Phosphatase, Balp in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.8ng/ml.. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05186; ALPL. Target Synonyms: AKP2; Alkaline phosphatase liver/bone/kidney; Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase tissue nonspecific isozyme; Alkaline phosphatase; tissue-nonspecific isozyme; Alkaline phosphomonoesterase; Alpl; AP TNAP; AP-TNAP; APTNAP; BAP; FLJ40094; FLJ93059; Glycerophosphatase; HOPS; Liver/bone/kidney type alkaline phosphatase; MGC161443; MGC167935; PHOA; PPBT_HUMAN; Tissue non specific alkaline phosphatase; Tissue nonspecific ALP; TNAP; TNSALP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7343