Learn More
Biomatik Corporation Mouse Regenerating Islet Derived Protein 3 Gamma (REG3g) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07048
Encompass_Preferred
Mouse Regenerating Islet Derived Protein 3 Gamma (REG3g) ELISA Kit for detecting REG3g in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O09049. Gene ID: 19695. Target Synonyms: REG-III; PAP1B; PAPIB; UNQ429; Pancreatitis-associated protein 1B; Regenerating islet-derived protein III-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3682