Learn More
Biomatik Corporation Human Retinol Binding Protein 1, Cellular (RBP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07101
Encompass_Preferred
Human Retinol Binding Protein 1, Cellular (RBP1) ELISA Kit for detecting RBP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09455. Gene ID: 5947. Target Synonyms: RBP-1; CRABP-I; CRBP; CRBP1; RBPC; Cellular retinol-binding protein I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3700