Learn More
Biomatik Corporation Human Platelet Factor 4, Pf-4 Elisa Kit
Supplier:  Biomatik Corporation EKC35046
Encompass_Preferred
Human Platelet Factor 4, Pf-4 Elisa Kit for detecting Platelet Factor 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02776; PF4. Target Synonyms: C-X-C motif chemokine 4; Chemokine (C X C motif) ligand 4; Chemokine (CXC motif) ligand 4; chemokine ligand 4; CXCL 4; CXCL4; Iroplact; MGC138298; Oncostatin A; Oncostatin-A; OncostatinA; PF 4; PF-4; PF4; Pf4a; Platelet factor 4; PLF4_HUMAN; SCYB 4; SCYB4; short form; Small inducible cytokine subfamily B; Small inducible cytokine subfamily member 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7520