Learn More
Biomatik Corporation Human Interleukin 37 (Il-37) Elisa Kit
Supplier:  Biomatik Corporation EKC41205
Encompass_Preferred
Human Interleukin 37 (Il-37) Elisa Kit for detecting Interleukin 1 Family, Member 7 (Zeta) in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NZH6; IL37. Target Synonyms: FIL1; FIL1 zeta; FIL1 (ZETA); FIL1Z; IL 1 zeta; IL 1F7; IL 1F7b (IL 1H4; IL 1H; IL 1RP1); IL 1H4; IL 1RP1; IL 1X; IL 1X protein; IL-1 zeta; IL-1F7; IL-1H; IL-1H4; IL-1RP1; IL-1X; IL-37; IL1F7 (canonical product IL 1F7b); IL1F7; IL1F7_HUMAN; IL1H4; IL1RP1; Interleukin 1 family member 7; interleukin 1 family; member 7 (zeta); Interleukin 1 homolog 4; Interleukin 1 related protein; Interleukin 1 superfamily z; Interleukin 1 zeta; Interleukin 37; Interleukin-1 family member 7; Interleukin-1 homolog 4; Interleukin-1 zeta; Interleukin-1-related protein; Interleukin-23. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7834