Learn More
Biomatik Corporation Human Sarcolipin (SLN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07194
Encompass_Preferred
Human Sarcolipin (SLN) ELISA Kit for detecting SLN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00631. Gene ID: 6588. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3741