Learn More
Biomatik Corporation Rat Selectin, Endothelium (SELE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07245
Encompass_Preferred
Rat E-selectin (E-selectin) ELISA Kit for detecting E-selectin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98105. Gene ID: 25544. Target Synonyms: CD62E; CD62-E; E-Selectin; ELAM; ELAM1; ESEL; SEL-E; E-LAM; E-LAM1; E-SEL; LECAM2; Endothelial Leukocyte Adhesion Molecule 1; CD62 Antigen-Like Family Member E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3763