Learn More
Biomatik Corporation Hamster TNF-α (Tumor Necrosis Factor-α) ELISA Kit
Supplier:  Biomatik Corporation EKF59542
Encompass_Preferred
Hamster TNF-α (Tumor Necrosis Factor-α) ELISA Kit for detecting TNF-α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Hamster. Detection Range: 3.125-200pg/ml. Sensitivity: 1.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A0A1U7RE37. Target Synonyms: TNF-α; DIF; TNF-alpha; TNFA; TNFSF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8272