Learn More
Biomatik Corporation Porcine TRY (Trypsin) ELISA Kit
Supplier:  Biomatik Corporation EKF59877
Encompass_Preferred
Porcine TRY (Trypsin) ELISA Kit for detecting Porcine TRY in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00761. Target Synonyms: Trypsin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8286