Learn More
Biomatik Corporation Human Serglycin (SRGN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07280
Encompass_Preferred
Human Serglycin (SRGN) ELISA Kit for detecting SRGN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10124. Gene ID: 5552. Target Synonyms: PRG; PPG; PRG1; proteoglycan 1, secretory granule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3785