Learn More
Biomatik Corporation Mouse Fgf15 (Fibroblast growth factor 15) ELISA Kit
Supplier:  Biomatik Corporation EKF60376
Encompass_Preferred
Mouse Fgf15 (Fibroblast growth factor 15) ELISA Kit for detecting Fgf15 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.688-300pg/ml. Sensitivity: 2.813pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35622. Target Synonyms: FGF15; Fibroblast growth factor 15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8313