Learn More
Biomatik Corporation Human Slit Homolog 1 (Slit1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07365
Encompass_Preferred
Human Slit Homolog 1 (Slit1) ELISA Kit for detecting Slit1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75093. Gene ID: 6585. Target Synonyms: MEGF4; SLIL1; SLIT3; Multiple epidermal growth factor-like domains protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3812