Learn More
Biomatik Corporation Mouse Stem Cell Factor (SCF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07464
Encompass_Preferred
Mouse Stem Cell Factor (SCF) ELISA Kit for detecting SCF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20826. Gene ID: 17311. Target Synonyms: KITLG; MGF; SF; KL1; Kitl; FPH2; Kit-Ligand; Steel Factor; Mast Cell Growth Factor; Steel factor; Familial Progressive Hyperpigmentation 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3839