Learn More
Biomatik Corporation Human Superoxide Dismutase 3, Extracellular (SOD3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07504
Encompass_Preferred
Human Superoxide Dismutase 3, Extracellular (SOD3) ELISA Kit for detecting SOD3 in serum, plasma, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8,000pg/ml. Sensitivity: 55pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08294. Target Synonyms: SOD-3; EC-SOD; Extracellular Superoxide Dismutase Cu-Zn. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3855