Learn More
Biomatik Corporation Human Suppressors of Cytokine Signaling 3 (SOCS3) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU07516
Human Suppressors Of Cytokine Signaling 3 (SOCS3) ELISA Kit for detecting SOCS3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14543. Gene ID: 9021. Target Synonyms: ATOD4; CIS3; Cish3; MGC71791; SOCS-3; SSI-3; SSI3; Cytokine-inducible SH2 protein 3; STAT-induced STAT inhibitor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.