Learn More
Biomatik Corporation Mouse Luteinizing Hormone (LH) ELISA Kit
Supplier:  Biomatik Corporation EKL61753
Encompass_Preferred
Mouse Luteinizing Hormone (LH) ELISA Kit for detecting LH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.468-30ng/ml. Sensitivity: 0.145ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Target Synonyms: ICSH; Lutropin; Interstitial Cell Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8866