Learn More
Biomatik Corporation MYT1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50413
Encompass_Preferred
MYT1 Colorimetric Cell-Based ELISA Kit for detecting MYT1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q99640. Gene ID: 9088. Target Synonyms: Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; Myt1 kinase; PKMYT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8951