Learn More
Biomatik Corporation Human Topoisomerase I (TOP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07766
Encompass_Preferred
Human Topoisomerase I (TOP1) ELISA Kit for detecting TOP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11387. Gene ID: 7150. Target Synonyms: TOPI; DNA Topoisomerase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3976