Learn More
Biomatik Corporation Mouse Transcription Factor A, Mitochondrial (TFAM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07779
Encompass_Preferred
Mouse Transcription Factor A, Mitochondrial (TFAM) ELISA Kit for detecting TFAM in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40630. Gene ID: 21780. Target Synonyms: MtTF1; TCF6; TCF6L2; mtTFA; Mitochondrial transcription factor 1; Transcription factor 6-like 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3980