Learn More
Biomatik Corporation Human Transferrin Receptor (TFR) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU07788
Human Transferrin Receptor (TFR) ELISA Kit for detecting TFR in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8,000pg/ml. Sensitivity: 57pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02786. Gene ID: 7037. Target Synonyms: CD71; TFR1; P90; TFRC; T9; Trfr; sTfR; Transferrin receptor protein 1, serum form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.