Learn More
Biomatik Corporation Mouse Transforming Growth Factor Beta Receptor II (TGFbR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07824
Encompass_Preferred
Mouse Transforming Growth Factor Beta Receptor II (TGFbR2) ELISA Kit for detecting TGFbR2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62312. Gene ID: 21813. Target Synonyms: MFS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4002