Learn More
Biomatik Corporation Human Trefoil Factor 1 (TFF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07869
Encompass_Preferred
Human Trefoil Factor 1 (TFF1) ELISA Kit for detecting TFF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04155. Gene ID: 7031. Target Synonyms: BCEI; HP1.A; HPS2; PS2; PNR2; Breast Cancer,Estrogen-Inducible Sequence Expressed In; Breast cancer estrogen-inducible protein; Polypeptide P1.A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4019