Learn More
Biomatik Corporation Pig Trypsin (TRY) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07915
Encompass_Preferred
Pig Trypsin (TRY) ELISA Kit for detecting TRY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00761. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4037