Learn More
Biomatik Corporation Human Tryptase (TPS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07922
Encompass_Preferred
Human Tryptase (TPS) ELISA Kit for detecting TPS in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15661-1. Gene ID: 7177. Target Synonyms: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4039