Learn More
Biomatik Corporation Human Interleukin 8 (IL8) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU08341
Encompass_Preferred
Human Interleukin 8 (IL8) High-sensitive ELISA Kit for detecting IL8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.9-250pg/ml. Sensitivity: 1.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: p10145. Gene ID: 3576. Target Synonyms: CXCL8; AMCF-I; GCP1; K60; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP1; SCYB8; TSG1; B-ENAP; Neutrophil-Activating Protein 1; Granulocyte Chemotactic Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6491