Learn More
Biomatik Corporation Human Laminin Alpha 2 (LAMa2) ELISA Kit
Supplier:  Biomatik Corporation EKU09104
Encompass_Preferred
Human Laminin Alpha 2 (LAMa2) ELISA Kit for detecting LAMa2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24043. Target Synonyms: LAM-A2; LAMM; Merosin; Congenital Muscular Dystrophy; Laminin M chain; Merosin heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6575