Learn More
Biomatik Corporation Human Wingless Type MMTV Integration Site Family, Member 7B (WNT7B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08219
Encompass_Preferred
Human Wingless Type MMTV Integration Site Family, Member 7B (WNT7B) ELISA Kit for detecting WNT7B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56706. Gene ID: 7477. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4158