Learn More
Biomatik Corporation Human WNT Inhibitory Factor 1 (WIF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08223
Encompass_Preferred
Human WNT Inhibitory Factor 1 (WIF1) ELISA Kit for detecting WIF1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5W5. Gene ID: 11197. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4159