Learn More
Biomatik Corporation Human Collagen Type III (COL3) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10529
Encompass_Preferred
Human Collagen Type III (COL3) Wide-range ELISA Kit for detecting COL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.264ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02461. Gene ID: 1281. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6701