Learn More
Biomatik Corporation Mouse Asprosin (Asprosin) ELISA Kit
Supplier:  Biomatik Corporation EKU10666
Encompass_Preferred
Mouse Asprosin (Asprosin) ELISA Kit for detecting Asprosin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61554. Gene ID: 14118. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6733