Learn More
Biomatik Corporation Human Dickkopf Related Protein 3 (DKK3) ELISA Kit
Supplier:  Biomatik Corporation EKU10792
Encompass_Preferred
Human Dickkopf Related Protein 3 (DKK3) ELISA Kit for detecting DKK3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBP4. Gene ID: 27122. Target Synonyms: REIC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6767