Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 15 (FGF15) ELISA Kit
Supplier:  Biomatik Corporation EKU10825
Encompass_Preferred
Mouse Fibroblast Growth Factor 15 (FGF15) ELISA Kit for detecting FGF15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-200pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O35622. Gene ID: 14170. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6774