Learn More
Biomatik Corporation Mouse Monoamine Oxidase A (MAOA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08424
Encompass_Preferred
Mouse Monoamine Oxidase A (MAOA) ELISA Kit for detecting MAOA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64133. Gene ID: 17161. Target Synonyms: MAO-A; Amine oxidase [flavin-containing] A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4217