Learn More
Biomatik Corporation Cattle Prolactin (PRL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08580
Encompass_Preferred
The Cattle Prolactin (PRL) ELISA Kit allows for quantitative determination of Prolactin in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 2.47-200 ng/mL. Sensitivity: 0.94 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01239. Cross Activity: No significant cross-reactivity or interference between Prolactin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4249