Learn More
Biomatik Corporation Human Amylase Alpha (Amya) ELISA Kit
Supplier:  Biomatik Corporation EKU02322
Encompass_Preferred
Human Amylase Alpha (Amya) ELISA Kit for detecting Amya in serum, plasma, saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04745. Gene ID: 276. Target Synonyms: Amy-a; 1,4-α-D-Glucan Glucanohydrolase; Glycogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5785