Learn More
Biomatik Corporation Human Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU08940
Encompass_Preferred
The Human Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit (CLIA) allows for quantitative determination of Neutrophil Gelatinase Associated Lipocalin in Serum, Plasma, Urine, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 27.4-20,000 pg/mL. Sensitivity: 9.9 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P80188. Cross Activity: No significant cross-reactivity or interference between Neutrophil Gelatinase Associated Lipocalin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4304