Learn More
Biomatik Corporation Mouse Hemopexin (HPX) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09099
Encompass_Preferred
Mouse Hemopexin (HPX) ELISA Kit for detecting HPX in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.44ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91X72. Gene ID: 15458. Target Synonyms: Haemopexin; Beta-1B-glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4343