Learn More
Biomatik Corporation Mouse Sortilin 1 (SORT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09140
Encompass_Preferred
Mouse Sortilin 1 (SORT1) ELISA Kit for detecting SORT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6PHU5. Gene ID: 20661. Target Synonyms: NT3; Gp95; NTR3; 100 kDa NT receptor; Glycoprotein 95; Neurotensin receptor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4355