Learn More
Biomatik Corporation Human Bisphosphoglycerate Mutase (BPGM) ELISA Kit
Supplier:  Biomatik Corporation EKU10680
Encompass_Preferred
Human Bisphosphoglycerate Mutase (BPGM) ELISA Kit for detecting BPGM in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07738. Gene ID: 669. Target Synonyms: 2,3-Bisphosphoglycerate Mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; BPG-dependent PGAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6736