Learn More
Biomatik Corporation Cattle Beta-Lactoglobulin (bLg) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09476
Encompass_Preferred
The Cattle Beta-Lactoglobulin (bLg) ELISA Kit (CLIA) allows for quantitative determination of Beta-Lactoglobulin in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 1.37-1,000 ng/mL. Sensitivity: 0.66 ng/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P02754. Cross Activity: No significant cross-reactivity or interference between Beta-Lactoglobulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4410