Learn More
Biomatik Corporation Human High Temperature Requirement Factor A1 (HTRA1) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09784
Encompass_Preferred
The Human High Temperature Requirement Factor A1 (HTRA1) ELISA Kit (CLIA) allows for quantitative determination of High Temperature Requirement Factor A1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 8.23-6,000 pg/mL. Sensitivity: 2.99 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: Q92743. Cross Activity: No significant cross-reactivity or interference between High Temperature Requirement Factor A1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4431