Learn More
Biomatik Corporation Human Troponin I Type 1, Slow Skeletal (TNNI1) ELISA Kit
Supplier:  Biomatik Corporation EKU07895
Encompass_Preferred
Human Troponin I Type 1, Slow Skeletal (TNNI1) ELISA Kit for detecting TNNI1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.88-3,000pg/ml. Sensitivity: 19.42pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19237. Gene ID: 7135. Target Synonyms: TNN1; Troponin I, slow-twitch isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6439