Learn More
Biomatik Corporation Human WNT Inhibitory Factor 1 (WIF1) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09989
Encompass_Preferred
Human WNT Inhibitory Factor 1 (WIF1) CLIA Kit for detecting WIF1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1000pg/ml. Sensitivity: 0.51pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9Y5W5. Gene ID: 11197. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4447