Learn More
Biomatik Corporation Mouse Substance P (SP) CLIA Kit
Supplier:  Biomatik Corporation EKU08616
Encompass_Preferred
Mouse Substance P (SP) CLIA Kit for detecting SP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 11.7-3,000pg/ml. Sensitivity: 4.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P41539. Gene ID: 21333. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6517